Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Titanium(IV) isopropoxide
Synonym(s):
Tetraisopropyl orthotitanate
Linear Formula:
Ti[OCH(CH
3
)
2
]
4
CAS Number:
546-68-9
Molecular Weight:
284.22
EC Number:
208-909-6
Products
Product Category
acids (3)
Brand
Sigma-Aldrich (5)
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Reagent Type
Core
Special Grade
Purum (1)
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
205273
Titanium(IV) isopropoxide,
97%
Expand
View Pricing
377996
Titanium(IV) isopropoxide,
99.999% trace metals basis
Expand
View Pricing
87560
Titanium(IV) isopropoxide,
≥97.0%
Expand
View Pricing
8.21895
Titanium(IV) isopropoxide,
Technipur
®
, for synthesis
Expand
View Pricing
687502
Titanium(IV) isopropoxide,
packaged for use in deposition systems
Expand
View Pricing