Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Titanium(IV) butoxide
Synonym(s):
Orthotitanic acid tetrabutylester, TNBT, TYZOR
®
TBT organic titanate, Tetrabutyl orthotitanate, Tetrabutyl titanate
Linear Formula:
Ti(OCH
2
CH
2
CH
2
CH
3
)
4
CAS Number:
5593-70-4
Molecular Weight:
340.32
EC Number:
227-006-8
Products
Brand
Sigma-Aldrich (2)
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Reagent Type
Core
Special Grade
Purum (1)
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
244112
Titanium(IV) butoxide,
reagent grade, 97%
Expand
View Pricing
86910
Titanium(IV) butoxide,
purum, ≥97.0% (gravimetric)
Expand
View Pricing