Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Titanium/Cobalt
Products
Manufacturer
Physical Form
Length (m)
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
GF04436399
Titanium/Cobalt,
wire, Ti 99.95%/Co 0.05%, 0.25 mm diameter, length 0.05 m, temper as drawn
Expand
View Pricing
GF45059485
Titanium/Cobalt,
wire, Ti 99.95%/Co 0.05%, 0.25 mm diameter, length 0.2 m, temper as drawn
Expand
View Pricing
GF00650360
Titanium/Cobalt,
wire, Ti 99.95%/Co 0.05%, 0.25 mm diameter, length 0.1 m, temper as drawn
Expand
View Pricing
GF02843456
Titanium/Cobalt,
wire, Ti 99.95%/Co 0.05%, 0.25 mm diameter, length 0.5 m, temper as drawn
Expand
View Pricing