Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Titanium carbide
Products
Manufacturer
Physical Form
Particle Size (µm)
Filter Products
Showing 1-8 of 8
Compare
Product Number
Description
Pricing
GF08821245
Titanium carbide,
powder, max. particle size 5 micron, weight 100 g, purity 99.8+%
Expand
View Pricing
GF14936092
Titanium carbide,
powder, mean particle size <0.05 micron, weight 100 g, purity 99.8+%
Expand
View Pricing
GF40873350
Titanium carbide,
powder, max. particle size 150 micron, weight 50 g, min. particle size 50 micron
Expand
View Pricing
GF87159101
Titanium carbide,
powder, 150 max. part. size (micron), weight 200 g, min. particle size 50 micron
Expand
View Pricing
GF23649037
Titanium carbide,
powder, mean particle size (fsss) 3 micron, weight 100 g, purity 98.5%
Expand
View Pricing
GF56129250
Titanium carbide,
powder, mean particle size (fsss) 3 micron, weight 200 g, purity 98.5%
Expand
View Pricing
GF91920269
Titanium carbide,
powder, 150 max. part. size (micron), weight 100 g, min. particle size 50 micron
Expand
View Pricing
GF47476637
Titanium carbide,
powder, mean particle size (fsss) 3 micron, weight 50 g, purity 98.5%
Expand
View Pricing