Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Thioglycolic acid solution
Synonym(s):
Mercaptoacetic acid
Linear Formula:
HSCH
2
COOH
CAS Number:
68-11-1
Molecular Weight:
92.12
EC Number:
200-677-4
Products
Brand
Sigma-Aldrich (1)
Supelco (1)
Formula Weight
Physical Form
Quality Segment
Special Grade
Purum (1)
Purum P.A. (1)
Technique
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
88652
Thioglycolic acid solution,
~80% in H
2
O, for spectrophotometric det. of palladium, iron, uranium(VI), molybdates and nitrites
Expand
View Pricing
T6750
Thioglycolic acid solution,
~70 % (w/w) in H
2
O
Expand
View Pricing