![Thiencarbazone-methyl PESTANAL®, analytical standard](/deepweb/assets/sigmaaldrich/product/structures/263/088/8b1db682-a184-444c-a9b7-7a2f51291d5e/640/8b1db682-a184-444c-a9b7-7a2f51291d5e.png)
Thiencarbazone-methyl
![Thiencarbazone-methyl PESTANAL®, analytical standard](/deepweb/assets/sigmaaldrich/product/structures/263/088/8b1db682-a184-444c-a9b7-7a2f51291d5e/640/8b1db682-a184-444c-a9b7-7a2f51291d5e.png)
Synonym(s):
Methyl 4-[(4,5-dihydro-3-methoxy-4-methyl-5-oxo-1H-1,2,4-triazol-1-yl)carbonylsulfamoyl]-5-methylthiophene-3-carboxylate
Empirical Formula (Hill Notation):
C12H14N4O7S2
CAS Number:
Molecular Weight:
390.39
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2