Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Thermal Transfer Tough-Tags
™
Products
Color
Manufacturer
Package Size
Filter Products
Showing 1-8 of 8
Compare
Product Number
Description
Pricing
Z743663
Thermal Transfer Tough-Tags
™
,
white, L × W 1 in. × 0.25 in., roll of 2000
Expand
View Pricing
Z743653
Thermal Transfer Tough-Tags
™
,
white, L × W 0.94 in. × 0.50 in., roll of 2000
Expand
View Pricing
Z743660
Thermal Transfer Tough-Tags
™
,
white, L × W 1.50 in. × 0.75 in., roll of 2000
Expand
View Pricing
Z743657
Thermal Transfer Tough-Tags
™
,
white, L × W 1.375 in. × 0.625 in., roll of 2000
Expand
View Pricing
Z743655
Thermal Transfer Tough-Tags
™
,
white, L × W 1.28 in. × 0.50 in., roll of 2000
Expand
View Pricing
Z743659
Thermal Transfer Tough-Tags
™
,
white, L × W 1.375 in. × 0.625 in., roll of 4000
Expand
View Pricing
Z743661
Thermal Transfer Tough-Tags
™
,
white, L × W 1.50 in. × 0.75 in., roll of 4000
Expand
View Pricing
Z743656
Thermal Transfer Tough-Tags
™
,
white, L × W 1.28 in. × 0.50 in., roll of 4000
Expand
View Pricing