Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Thermal Transfer Cryo-Tags
®
Products
Color
Manufacturer
Length (in)
Technique
Filter Products
Showing 1-17 of 17
Compare
Product Number
Description
Pricing
Z743449
Thermal Transfer Cryo-Tags
®
,
white, L × W 1.05 in. × 0.50 in., pkg of 2000 labels Roll
Expand
View Pricing
Z743455
Thermal Transfer Cryo-Tags
®
,
white, L × W 1.375 in. × 0.625 in., pkg of 2000 labels Roll
Expand
View Pricing
Z743458
Thermal Transfer Cryo-Tags
®
,
white, L × W 1.50 in. × 0.50 in., pkg of 2000 labels Roll
Expand
View Pricing
Z743452
Thermal Transfer Cryo-Tags
®
,
white, L × W 1.28 in. × 0.50 in., pkg of 4000 labels Roll
Expand
View Pricing
Z743447
Thermal Transfer Cryo-Tags
®
,
white, L × W 0.94 in. × 0.50 in., pkg of 2000 labels Roll
Expand
View Pricing
Z743454
Thermal Transfer Cryo-Tags
®
,
white, L × W 1.375 in. × 0.625 in., pkg of 4000 labels Roll
Expand
View Pricing
Z743448
Thermal Transfer Cryo-Tags
®
,
white, L × W 0.94 in. × 0.50 in., pkg of 4000 labels Roll
Expand
View Pricing
Z743456
Thermal Transfer Cryo-Tags
®
,
white, L × W 1.5 in. × 0.75 in., pkg of 2000 labels Roll
Expand
View Pricing
Z743462
Thermal Transfer Cryo-Tags
®
,
white, L × W 2.0 in. × 0.25 in., pkg of 2000 labels Roll
Expand
View Pricing
Z743446
Thermal Transfer Cryo-Tags
®
,
white, L × W 0.81 in. × 0.28 in., pkg of 4000 labels Roll
Expand
View Pricing
Z743463
Thermal Transfer Cryo-Tags
®
,
white, L × W 2.0 in. × 0.25 in., pkg of 4000 labels Roll
Expand
View Pricing
Z743444
Thermal Transfer Cryo-Tags
®
,
white, L × W 0.81 in. × 0.28 in., pkg of 2000 labels Roll
Expand
View Pricing
Z743451
Thermal Transfer Cryo-Tags
®
,
white, L × W 1.28 in. × 0.50 in., pkg of 2000 labels Roll
Expand
View Pricing
Z744099
Thermal Transfer Cryo-Tags
®
,
white, W × L 3.0 in. × 2.0 in., pack of 1000 ea (labels per roll)
Expand
View Pricing
Z743457
Thermal Transfer Cryo-Tags
®
,
white, L × W 1.5 in. × 0.75 in., pkg of 4000 labels Roll
Expand
View Pricing
Z743459
Thermal Transfer Cryo-Tags
®
,
white, L × W 1.50 in. × 0.50 in., pkg of 4000 labels Roll
Expand
View Pricing
Z743460
Thermal Transfer Cryo-Tags
®
,
white, L × W 1.50 in. × 0.75 in., pkg of 4000 labels Roll
Expand
View Pricing