Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
ThalesNano CatCart
®
catalyst cartridge system, 70 mm L
Products
Sustainability
Feature
Manufacturer
Diameter (mm)
Package Size
Filter Products
Showing 1-20 of 65
Page 1 of 4
Page 1 of 4
Compare
Product Number
Description
Pricing
THS01131
ThalesNano CatCart
®
catalyst cartridge system, 70 mm L,
10% Pd/C
Expand
View Pricing
THS01133
ThalesNano CatCart
®
catalyst cartridge system, 70 mm L,
10%Pt/C
Expand
View Pricing
THS05134
ThalesNano CatCart
®
catalyst cartridge system, 70 mm L,
tetrakis (TPP)-palladium
Expand
View Pricing
THS08132
ThalesNano CatCart
®
catalyst cartridge system, 70 mm L,
5% Pt/C
Expand
View Pricing
THS05131
ThalesNano CatCart
®
catalyst cartridge system, 70 mm L,
Pd EnCat
®
30
Expand
View Pricing
THS02131
ThalesNano CatCart
®
catalyst cartridge system, 70 mm L,
5% Pd/C
Expand
View Pricing
THS04135
ThalesNano CatCart
®
catalyst cartridge system, 70 mm L,
1% Pd/C
Expand
View Pricing
THS01135
ThalesNano CatCart
®
catalyst cartridge system, 70 mm L,
20%Pd(OH)
2
/C
Expand
View Pricing
THS01132
ThalesNano CatCart
®
catalyst cartridge system, 70 mm L,
Raney nickel
Expand
View Pricing
THS01030
ThalesNano CatCart
®
catalyst cartridge system, 70 mm L,
inert quartz
Expand
View Pricing
THS01639
ThalesNano CatCart
®
catalyst cartridge system, 70 mm L,
Au/TiO
2
Expand
View Pricing
THS03138
ThalesNano CatCart
®
catalyst cartridge system, 70 mm L,
1% Pd/Al
2
O
3
Expand
View Pricing
THS01531
ThalesNano CatCart
®
catalyst cartridge system, 70 mm L,
10% Pd/CaCO
3
Expand
View Pricing
THS05137
ThalesNano CatCart
®
catalyst cartridge system, 70 mm L,
Wilkinson′s catalyst RhCl(TPP)
3
Expand
View Pricing
THS01439
ThalesNano CatCart
®
catalyst cartridge system, 70 mm L,
1% Pt/C, doped with V
Expand
View Pricing
THS01136
ThalesNano CatCart
®
catalyst cartridge system, 70 mm L,
Raney cobalt
Expand
View Pricing
THS01134
ThalesNano CatCart
®
catalyst cartridge system, 70 mm L,
5% Rh/C
Expand
View Pricing
THS04133
ThalesNano CatCart
®
catalyst cartridge system, 70 mm L,
Ni/SiO
2
-Al
2
O
3
Expand
View Pricing
THS01138
ThalesNano CatCart
®
catalyst cartridge system, 70 mm L,
5% Pd/Al
2
O
3
Expand
View Pricing
THS03135
ThalesNano CatCart
®
catalyst cartridge system, 70 mm L,
5% Pd (+1%Fe)/C
Expand
View Pricing
Page 1 of 4
Page 1 of 4