Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
ThalesNano CatCart
®
catalyst cartridge system, 30 mm L
EC Number:
231-115-6
Products
Manufacturer
Diameter (mm)
Package Size
Filter Products
Showing 1-20 of 57
Page 1 of 3
Page 1 of 3
Compare
Product Number
Description
Pricing
THS01111
ThalesNano CatCart
®
catalyst cartridge system, 30 mm L,
10% Pd/C
Expand
View Pricing
THS01115
ThalesNano CatCart
®
catalyst cartridge system, 30 mm L,
20%Pd(OH)
2
/C
Expand
View Pricing
THS05117
ThalesNano CatCart
®
catalyst cartridge system, 30 mm L,
Wilkinson′s catalyst RhCl(TPP)
3
Expand
View Pricing
THS02111
ThalesNano CatCart
®
catalyst cartridge system, 30 mm L,
5% Pd/C
Expand
View Pricing
THS01113
ThalesNano CatCart
®
catalyst cartridge system, 30 mm L,
10% Pt/C
Expand
View Pricing
THS05114
ThalesNano CatCart
®
catalyst cartridge system, 30 mm L,
tetrakis (TPP)-palladium
Expand
View Pricing
THS01118
ThalesNano CatCart
®
catalyst cartridge system, 30 mm L,
5% Pd/Al
2
O
3
Expand
View Pricing
THS01114
ThalesNano CatCart
®
catalyst cartridge system, 30 mm L,
5% Rh/C
Expand
View Pricing
THS01110
ThalesNano CatCart
®
catalyst cartridge system, 30 mm L,
inert titanium
Expand
View Pricing
THS05111
ThalesNano CatCart
®
catalyst cartridge system, 30 mm L,
Pd EnCat
®
30
Expand
View Pricing
THS01319
ThalesNano CatCart
®
catalyst cartridge system, 30 mm L,
Quadrasil
®
AP
Expand
View Pricing
THS03118
ThalesNano CatCart
®
catalyst cartridge system, 30 mm L,
1% Pd/Al
2
O
3
Expand
View Pricing
THS02114
ThalesNano CatCart
®
catalyst cartridge system, 30 mm L,
5% Pd/CaCO
3
/Pb
Expand
View Pricing
THS01116
ThalesNano CatCart
®
catalyst cartridge system, 30 mm L,
Raney cobalt
Expand
View Pricing
THS01419
ThalesNano CatCart
®
catalyst cartridge system, 30 mm L,
1% Pt/C, doped with V
Expand
View Pricing
THS01312
ThalesNano CatCart
®
catalyst cartridge system, 30 mm L,
triamine-tetraacetate-functionalized silica
Expand
View Pricing
THS01322
ThalesNano CatCart
®
catalyst cartridge system, 30 mm L,
Quadrasil
®
MP
Expand
View Pricing
THS01318
ThalesNano CatCart
®
catalyst cartridge system, 30 mm L,
QuadraPure
®
MPA
Expand
View Pricing
THS01615
ThalesNano CatCart
®
catalyst cartridge system, 30 mm L,
Rh(COD)(dppf)/SiO
2
Expand
View Pricing
THS03119
ThalesNano CatCart
®
catalyst cartridge system, 30 mm L,
10% Pd/Al
2
O
3
Expand
View Pricing
Page 1 of 3
Page 1 of 3