Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Tetramethylammonium chloride
Synonym(s):
TMA, Tetramethylammonium chloride
Linear Formula:
(CH
3
)
4
N(Cl)
CAS Number:
75-57-0
Molecular Weight:
109.60
EC Number:
200-880-8
Products
Brand
Sigma-Aldrich (1)
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Technique
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
87718
Tetramethylammonium chloride,
BioUltra, Molecular Biology, ≥99.0% (AT)
Expand
View Pricing
T19526
Tetramethylammonium chloride,
reagent grade, ≥98%
Expand
View Pricing
8.22156
Tetramethylammonium chloride,
for synthesis
Expand
View Pricing
74202
Tetramethylammonium chloride,
suitable for ion pair chromatography, LiChropur
™
, ≥99.0% (AT)
Expand
View Pricing