![Tazobactam sodium salt analytical standard](/deepweb/assets/sigmaaldrich/product/structures/126/662/bfc30f9a-9952-4537-90d3-26c7de1d4bd0/640/bfc30f9a-9952-4537-90d3-26c7de1d4bd0.png)
Tazobactam sodium salt
![Tazobactam sodium salt analytical standard](/deepweb/assets/sigmaaldrich/product/structures/126/662/bfc30f9a-9952-4537-90d3-26c7de1d4bd0/640/bfc30f9a-9952-4537-90d3-26c7de1d4bd0.png)
Synonym(s):
CL-298741, YTR-830H, [2S-(2α, 3β, 5α)]-3-Methyl-7-oxo-3(1H,2,3-triazol-1-ylmethyl)-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid 4,4-dioxide
Empirical Formula (Hill Notation):
C10H11N4NaO5S
CAS Number:
Molecular Weight:
322.27
Products
Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Activity Spectrum
Format
Quality Segment
Application
Mode of Action
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing