Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Tantalum(V) chloride
Synonym(s):
Tantalum pentachloride
Linear Formula:
TaCl
5
CAS Number:
7721-01-9
Molecular Weight:
358.21
EC Number:
231-755-6
Products
Brand
Sigma-Aldrich (3)
Formula Weight
Physical Form
Purity
Reagent Type
Core
Special Grade
Anhydrous (1)
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
400475
Tantalum(V) chloride,
99.99% trace metals basis
Expand
View Pricing
218634
Tantalum(V) chloride,
99.8% trace metals basis
Expand
View Pricing
510688
Tantalum(V) chloride,
anhydrous, powder, 99.999% trace metals basis
Expand
View Pricing