
Tamsulosin hydrochloride

Synonym(s):
5-[(2R)-2-[[2-(2-Ethoxyphenoxy)ethyl]amino]propyl]-2-methoxybenzenesulfonamide hydrochloride, Tamsulosin hydrochloride
Empirical Formula (Hill Notation):
C20H28N2O5S · HCl
CAS Number:
Molecular Weight:
444.97
Products
Product Category
Brand
Color
Formula Weight
Gene Alias
Manufacturer
Physical Form
Purity
Agency
Format
Research Area
Quality Segment
Application
Package Size
API Family
Special Grade
Technique
Showing 1-5 of 5
Compare
Product Number
Description
Pricing