
Tacrolimus for system suitability

Synonym(s):
FK-506 monohydrate, Tacrolimus
Empirical Formula (Hill Notation):
C44H69NO12 · H2O
CAS Number:
Molecular Weight:
822.03
Products
Formula Weight
Gene Alias
Manufacturer
Format
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing