Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Syringes Fitted with Luer Lock Valve (509353)
Products
Product Category
syringes (5)
Manufacturer
Volume (ml)
Volume (µL)
Package Size
Gauge
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
509655
Syringes Fitted with Luer Lock Valve (509353),
volume 100 mL, 100MR-VLLMA-GT, needle size (not included)
Expand
View Pricing
509612
Syringes Fitted with Luer Lock Valve (509353),
volume 5 mL, 5MDR-VLLMA-GT
Expand
View Pricing
509620
Syringes Fitted with Luer Lock Valve (509353),
volume 10 mL, 10MDR-VLLMA-GT, needle size (not included)
Expand
View Pricing
509639
Syringes Fitted with Luer Lock Valve (509353),
volume 25 mL, 25MDR-VLLMA-GT, needle size (not included)
Expand
View Pricing
509590
Syringes Fitted with Luer Lock Valve (509353),
volume 1000 μL, 1MDR-VLL-GT, needle size (not included)
Expand
View Pricing