Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Synthware
™
two neck round bottom flask with angled side neck
Products
Product Category
flasks (23)
Manufacturer
Capacity (mL)
Filter Products
Showing 1-20 of 23
Page 1 of 2
Page 1 of 2
Compare
Product Number
Description
Pricing
SYNF414450
Synthware
™
two neck round bottom flask with angled side neck,
50 mL, center joint: ST/NS 24/40, side joint: ST/NS 24/40
Expand
View Pricing
SYNF41241L
Synthware
™
two neck round bottom flask with angled side neck,
1000 mL, center joint: ST/NS 24/40, side joint: ST/NS 24/40
Expand
View Pricing
SYNF414100
Synthware
™
two neck round bottom flask with angled side neck,
100 mL, center joint: ST/NS 24/40, side joint: ST/NS 14/20
Expand
View Pricing
SYNF411110
Synthware
™
two neck round bottom flask with angled side neck,
10 mL, center joint: ST/NS 14/20, side joint: ST/NS 14/20
Expand
View Pricing
SYNF411100
Synthware
™
two neck round bottom flask with angled side neck,
100 mL, center joint: ST/NS 14/20, side joint: ST/NS 14/20
Expand
View Pricing
SYNF414400
Synthware
™
two neck round bottom flask with angled side neck,
100 mL, center joint: ST/NS 24/40, side joint: ST/NS 24/40
Expand
View Pricing
SYNF41611L
Synthware
™
two neck round bottom flask with angled side neck,
1000 mL, center joint: ST/NS 24/40, side joint: ST/NS 14/20
Expand
View Pricing
SYNF411250
Synthware
™
two neck round bottom flask with angled side neck,
250 mL, center joint: ST/NS 14/20, side joint: ST/NS 14/20
Expand
View Pricing
SYNF414150
Synthware
™
two neck round bottom flask with angled side neck,
50 mL, center joint: ST/NS 24/40, side joint: ST/NS 14/20
Expand
View Pricing
SYNF411125
Synthware
™
two neck round bottom flask with angled side neck,
25 mL, center joint: ST/NS 14/20, side joint: ST/NS 14/20
Expand
View Pricing
SYNF411150
Synthware
™
two neck round bottom flask with angled side neck,
50 mL, center joint: ST/NS 14/20, side joint: ST/NS 14/20
Expand
View Pricing
SYNF419950
Synthware
™
two neck round bottom flask with angled side neck,
50 mL, center joint: ST/NS 19/22, side joint: ST/NS 19/22
Expand
View Pricing
SYNF414501
Synthware
™
two neck round bottom flask with angled side neck,
500 mL, center joint: ST/NS 24/40, side joint: ST/NS 14/20
Expand
View Pricing
SYNF419900
Synthware
™
two neck round bottom flask with angled side neck,
100 mL, center joint: ST/NS 19/22, side joint: ST/NS 19/22
Expand
View Pricing
SYNF414251
Synthware
™
two neck round bottom flask with angled side neck,
250 mL, center joint: ST/NS 24/40, side joint: ST/NS 14/20
Expand
View Pricing
SYNF414504
Synthware
™
two neck round bottom flask with angled side neck,
500 mL, center joint: ST/NS 24/40, side joint: ST/NS 24/40
Expand
View Pricing
SYNF414250
Synthware
™
two neck round bottom flask with angled side neck,
250 mL, center joint: ST/NS 24/40, side joint: ST/NS 24/40
Expand
View Pricing
SYNF419100
Synthware
™
two neck round bottom flask with angled side neck,
100 mL, center joint: ST/NS 24/40, side joint: ST/NS 19/22
Expand
View Pricing
SYNF410250
Synthware
™
two neck round bottom flask with angled side neck,
250 mL, center joint: ST/NS 19/22, side joint: ST/NS 14/20
Expand
View Pricing
SYNF411501
Synthware
™
two neck round bottom flask with angled side neck,
500 mL, center joint: ST/NS 14/20, side joint: ST/NS 14/20
Expand
View Pricing
Page 1 of 2
Page 1 of 2