Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Synthware
™
three neck round bottom flask with vertical side necks
Products
Product Category
flasks (19)
Manufacturer
Capacity (mL)
Filter Products
Showing 1-19 of 19
Compare
Product Number
Description
Pricing
SYNF444000
Synthware
™
three neck round bottom flask with vertical side necks,
1000 mL, center joint: ST/NS 24/40, side joint: ST/NS 24/40
Expand
View Pricing
SYNF444250
Synthware
™
three neck round bottom flask with vertical side necks,
250 mL, center joint: ST/NS 24/40, side joint: ST/NS 24/40
Expand
View Pricing
SYNF44243L
Synthware
™
three neck round bottom flask with vertical side necks,
bottle capacity 3000 mL, side joint: ST/NS 24/40
Expand
View Pricing
SYNF444500
Synthware
™
three neck round bottom flask with vertical side necks,
500 mL, center joint: ST/NS 24/40, side joint: ST/NS 24/40
Expand
View Pricing
SYNF444900
Synthware
™
three neck round bottom flask with vertical side necks,
100 mL, center joint: ST/NS 24/40, side joint: ST/NS 19/22
Expand
View Pricing
SYNF44242L
Synthware
™
three neck round bottom flask with vertical side necks,
2000 mL, center joint: ST/NS 24/40, side joint: ST/NS 24/40
Expand
View Pricing
SYNF44841L
Synthware
™
three neck round bottom flask with vertical side necks,
1000 mL, center joint: ST/NS 29/42, side joint: ST/NS 24/40
Expand
View Pricing
SYNF44542L
Synthware
™
three neck round bottom flask with vertical side necks,
2000 mL, center joint: ST/NS 45/50, side joint: ST/NS 24/40
Expand
View Pricing
SYNF449900
Synthware
™
three neck round bottom flask with vertical side necks,
100 mL, center joint: ST/NS 19/22, side joint: ST/NS 19/22
Expand
View Pricing
SYNF441100
Synthware
™
three neck round bottom flask with vertical side necks,
100 mL, center joint: ST/NS 14/20, side joint: ST/NS 14/20
Expand
View Pricing
SYNF444400
Synthware
™
three neck round bottom flask with vertical side necks,
100 mL, center joint: ST/NS 24/40, side joint: ST/NS 24/40
Expand
View Pricing
SYNF441150
Synthware
™
three neck round bottom flask with vertical side necks,
50 mL, center joint: ST/NS 14/20, side joint: ST/NS 14/20
Expand
View Pricing
SYNF442250
Synthware
™
three neck round bottom flask with vertical side necks,
250 mL, center joint: ST/NS 24/40, side joint: ST/NS 19/22
Expand
View Pricing
SYNF446500
Synthware
™
three neck round bottom flask with vertical side necks,
500 mL, center joint: ST/NS 29/42, side joint: ST/NS 24/40
Expand
View Pricing
SYNF441250
Synthware
™
three neck round bottom flask with vertical side necks,
250 mL, center joint: ST/NS 14/20, side joint: ST/NS 14/20
Expand
View Pricing
SYNF44842L
Synthware
™
three neck round bottom flask with vertical side necks,
2000 mL, center joint: ST/NS 29/42, side joint: ST/NS 24/40
Expand
View Pricing
SYNF44292L
Synthware
™
three neck round bottom flask with vertical side necks,
2000 mL, center joint: ST/NS 29/42, side joint: ST/NS 29/42
Expand
View Pricing
SYNF448500
Synthware
™
three neck round bottom flask with vertical side necks,
500 mL, center joint: ST/NS 29/42, side joint: ST/NS 29/42
Expand
View Pricing
SYNF441500
Synthware
™
three neck round bottom flask with vertical side necks,
500 mL, center joint: ST/NS 14/20, side joint: ST/NS 14/20
Expand
View Pricing