Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Synthware
™
solvent still head
Products
Manufacturer
Filter Products
Showing 1-11 of 11
Compare
Product Number
Description
Pricing
SYNS251250
Synthware
™
solvent still head,
250 mL, joint: ST/NS 14/20
Expand
View Pricing
SYNS244004
Synthware
™
solvent still head,
1000 mL, top joint: ST/NS 24/40, bottom joint: ST/NS 24/40
Expand
View Pricing
SYNS254250
Synthware
™
solvent still head,
250 mL, joint: ST/NS 24/40, side joint: 14/20
Expand
View Pricing
SYNS244504
Synthware
™
solvent still head,
500 mL, top joint: ST/NS 24/40, bottom joint: ST/NS 24/40
Expand
View Pricing
SYNS234504
Synthware
™
solvent still head,
500 mL, top joint: ST/NS 24/40, bottom joint: ST/NS 24/40, side joint: 14/20
Expand
View Pricing
SYNS234004
Synthware
™
solvent still head,
1000 mL, top joint: ST/NS 24/40, bottom joint: ST/NS 24/40, side joint: 14/20
Expand
View Pricing
SYNS254500
Synthware
™
solvent still head,
500 mL, joint: ST/NS 24/40, side joint: 14/20
Expand
View Pricing
SYNS144125
Synthware
™
solvent still head,
125 mL, bottom joint: ST/NS 24/40
Expand
View Pricing
SYNS134250
Synthware
™
solvent still head,
250 mL, joint: ST/NS 24/40, side joint: ST/NS 14/10, side joint: 14/20
Expand
View Pricing
SYNS134500
Synthware
™
solvent still head,
500 mL, joint: ST/NS 24/40, side joint: ST/NS 14/10, side joint: 14/20
Expand
View Pricing
SYNS254000
Synthware
™
solvent still head,
1000 mL, joint: ST/NS 24/40, side joint: 14/20
Expand
View Pricing