Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Synthware
™
reaction tube with glass stopcock
Products
Product Category
tubes (8)
Manufacturer
Capacity (mL)
Color
Package Size
Sterility
Filter Products
Showing 1-8 of 8
Compare
Product Number
Description
Pricing
SYNF891410
Synthware
™
reaction tube with glass stopcock,
10 mL, joint: ST/NS 14/20
Expand
View Pricing
SYNF891425
Synthware
™
reaction tube with glass stopcock,
25 mL, joint: ST/NS 14/20
Expand
View Pricing
SYNF891950
Synthware
™
reaction tube with glass stopcock,
50 mL, joint: ST/NS 19/22
Expand
View Pricing
SYNF891450
Synthware
™
reaction tube with glass stopcock,
50 mL, joint: ST/NS 14/20
Expand
View Pricing
SYNF891100
Synthware
™
reaction tube with glass stopcock,
100 mL, joint: ST/NS 14/20
Expand
View Pricing
SYNF894100
Synthware
™
reaction tube with glass stopcock,
100 mL, joint: ST/NS 24/40
Expand
View Pricing
SYNF892450
Synthware
™
reaction tube with glass stopcock,
50 mL, joint: ST/NS 24/40
Expand
View Pricing
SYNF894250
Synthware
™
reaction tube with glass stopcock,
250 mL, joint: ST/NS 24/40
Expand
View Pricing