Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Swabs and SwabChecks
Products
Product Category
swabs (7)
Manufacturer
Capacity (mL)
Application
Microorganism Suitability
Package Size
Filter Products
Showing 1-7 of 7
Compare
Product Number
Description
Pricing
Z699381
Swabs and SwabChecks,
Total Count Swab Kit
Expand
View Pricing
Z699446
Swabs and SwabChecks,
Hygiene SwabCheck
Expand
View Pricing
Z699454
Swabs and SwabChecks,
Listeria SwabCheck
Expand
View Pricing
Z699365
Swabs and SwabChecks,
Buffer Swabs, capacity 4 mL
Expand
View Pricing
Z699497
Swabs and SwabChecks,
Polywipe Sponge
Expand
View Pricing
Z699403
Swabs and SwabChecks,
Yeast & Mold Swab Kit
Expand
View Pricing
Z699438
Swabs and SwabChecks,
Coliform SwabCheck
Expand
View Pricing