Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Supelclean
™
ENVI-Florisil
®
SPE Tubes
Products
Product Category
tubes (4)
Brand
Supelco (4)
Product Line
Agency
Chemical Composition
Separation Technique
Volume (ml)
Application
Package Size
Technique
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
57046
Supelclean
™
ENVI-Florisil
®
SPE Tubes,
stainless steel frit, bed wt. 500 mg, volume 6 mL, pk of 30
Expand
View Pricing
57053
Supelclean
™
ENVI-Florisil
®
SPE Tubes,
stainless steel frit, bed wt. 1 g, volume 6 mL, pk of 30
Expand
View Pricing
57058
Supelclean
™
ENVI-Florisil
®
SPE Tubes,
PTFE frit, bed wt. 500 mg, volume 3 mL, pk of 54
Expand
View Pricing
54095-U
Supelclean
™
ENVI-Florisil
®
SPE Tubes,
glass hardware, PTFE frit, bed wt. 1 g, volume 6 mL, pk of 30
Expand
View Pricing