Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Supelclean
™
ENVI-Carb
™
II/PSA SPE Tube
Products
Product Category
tubes (5)
Brand
Supelco (1)
Product Line
Chemical Composition
Separation Technique
Volume (ml)
Application
Package Size
Technique
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
54217-U
Supelclean
™
ENVI-Carb
™
II/PSA SPE Tube,
bed A 500 mg (Supelclean
™
ENVI-Carb-II SPE), bed B 500 mg (Supelclean
™
PSA SPE), volume 20 mL, pk of 20
Expand
View Pricing
54067-U
Supelclean
™
ENVI-Carb
™
II/PSA SPE Tube,
bed B 500 mg (Supelclean
™
PSA SPE), bed A 500 mg (Supeclean
™
ENVI-Carb-II SPE), volume 6 mL, pk of 30
Expand
View Pricing
54103-U
Supelclean
™
ENVI-Carb
™
II/PSA SPE Tube,
bed B 500 mg (Supelclean
™
PSA SPE), bed A 500 mg (Supelclean
™
ENVI-Carb-II SPE), volume 6 mL, pk of 300
Expand
View Pricing
54058-U
Supelclean
™
ENVI-Carb
™
II/PSA SPE Tube,
bed B 600 mg (Supelclean
™
PSA SPE), bed A 300 mg (Supelclean
™
ENVI-Carb-II SPE), volume 6 mL, pk of 30
Expand
View Pricing
55119-U
Supelclean
™
ENVI-Carb
™
II/PSA SPE Tube,
bed A 500 mg (Supelclean
™
Envi-Carb-II SPE), bed B 300 mg (Supelclean
™
PSA SPE), volume 6 mL, pk of 30
Expand
View Pricing