<iframe src="https://www.googletagmanager.com/ns.html?id=GTM-TXMJ2GG>m_auth=>m_preview=>m_cookies_win=x" height="0" width="0" style="display:none;visibility:hidden" id="tag-manager"></iframe>
Skip to Content
Products
Cart
0
AE
EN
Products
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Login
Back
Sulfuric acid solution
Synonym(s):
Sulfuric acid solution
Linear Formula:
H
2
SO
4
CAS Number:
7664-93-9
Molecular Weight:
98.08
Products
Product Category
acids (8)
inorganic acids (1)
reagents (1)
Brand
Sigma-Aldrich (1)
Supelco (10)
Formula Weight
Physical Form
Product Line
Agency
Quality Segment
Package Size
Reaction Type
Special Grade
Puriss (1)
Puriss P.A. (1)
Technique
Filter Products
Showing 1-12 of 12
Compare
Product Number
Description
Pricing
1.09072
Sulfuric acid solution,
c(H₂SO₄) = 0.5 mol/l (1 N), reag. Ph. Eur., reag. USP, ready-to-use volumetric solution for titration, Titripur
®
Expand
View Pricing
1.09074
Sulfuric acid solution,
c(H₂SO₄) = 0.05 mol/l (0.1 N), reag. USP, ready-to-use volumetric solution for titration, Titripur
®
Expand
View Pricing
1.60313
Sulfuric acid solution,
c(H₂SO₄) = 1 mol/l (2 N), ready-to-use volumetric solution for titration, Titripur
®
Expand
View Pricing
1.09981
Sulfuric acid solution,
c(H₂SO₄) = 0.5 mol/l (1 N), concentrate for preparing 1000 ml volumetric solution for titration, Titrisol
®
Expand
View Pricing
1.60315
Sulfuric acid solution,
c(H₂SO₄), 5 M (10 N), ready-to-use volumetric solution for titration, Titripur
®
Expand
View Pricing
84736
Sulfuric acid solution,
puriss. p.a., ≥25% (T)
Expand
View Pricing
1.09984
Sulfuric acid solution,
c(H₂SO₄) = 0.05 mol/l (0.1 N), concentrate for preparing 1000 ml volumetric solution for titration, Titrisol
®
Expand
View Pricing
1.09073
Sulfuric acid solution,
c(H₂SO₄) 0.25 mol/l (0.5 N), ready-to-use volumetric solution for titration, Titripur
®
Expand
View Pricing
4.80364
Sulfuric acid solution,
c(H₂SO₄) = 2.5 mol/l (5 N), ready-to-use volumetric solution for titration, Titripur
®
Expand
View Pricing
1.60314
Sulfuric acid solution,
c(H₂SO₄) = 0.005 mol/l (0.01 N), reag. USP, ready-to-use volumetric solution for titration, Titripur
®
Expand
View Pricing
1.09982
Sulfuric acid solution,
c(H₂SO₄) = 0.005 mol/l (0.01 N), concentrate for preparing 1000 ml volumetric solution for titration, Titrisol
®
Expand
View Pricing
1.09912
Sulfuric acid solution,
c(H₂SO₄) = 2.5 mol/l (5 N), 1 L solution for the preparation of dilute volumetric solutions or for direct use, Combi-Titrisol
®
Expand
View Pricing
Choose up to 4 products to compare
Compare
Clear All
English - EN
Learn More