Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Sulfatase from
Helix pomatia
Synonym(s):
Aryl-sulfatase, Aryl-sulfate sulfohydrolase, Phenolsulfatase
CAS Number:
9016-17-5
Products
Brand
Sigma-Aldrich (2)
Physical Form
Application
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
S9626
Sulfatase from
Helix pomatia
,
Type H-1, sulfatase ≥10,000 units/g solid
Expand
View Pricing
S9751
Sulfatase from
Helix pomatia
,
Type H-2, aqueous solution, ≥2,000 units/mL
Expand
View Pricing