Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Strep-Tactin XT Sepharose chromatography resin
Synonym(s):
StrepTrap
™
High Performance
Products
Product Category
resins (2)
Brand
Sigma-Aldrich (2)
Manufacturer
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
GE29401324
Strep-Tactin XT Sepharose chromatography resin,
pkg of 1 × 10 mL, chromatography resin
Expand
View Pricing
GE29401326
Strep-Tactin XT Sepharose chromatography resin,
pkg of 1 × 50 mL, chromatography resin
Expand
View Pricing