Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Storage Box for Eppendorf
®
Tubes
Products
Product Category
boxes (9)
Manufacturer
Height (mm)
Package Size
Filter Products
Showing 1-9 of 9
Compare
Product Number
Description
Pricing
EP0030140613
Storage Box for Eppendorf
®
Tubes,
size 5 × 5, pack of 2 ×, Holds 25 x 5 mL tubes
Expand
View Pricing
EP0030140532
Storage Box for Eppendorf
®
Tubes,
size 5 × 5, pack of 4 ×, Holds 25 x 5 mL tubes
Expand
View Pricing
EP0030140524
Storage Box for Eppendorf
®
Tubes,
size 8 × 8, pack of 3 ×, Holds 64 x 1-2 mL tubes
Expand
View Pricing
EP0030140567
Storage Box for Eppendorf
®
Tubes,
size 9 × 9, Holds 81 x 4-5 mL tubes
Expand
View Pricing
EP0030140583
Storage Box for Eppendorf
®
Tubes,
size 5 × 5, pack of 2 ×, Holds 25 x 15 mL tubes
Expand
View Pricing
EP0030140591
Storage Box for Eppendorf
®
Tubes,
size 3 × 3, pack of 2 ×, Holds 9 x 50 mL tubes, Holds 4 x 15 mL tubes
Expand
View Pricing
EP0030140508
Storage Box for Eppendorf
®
Tubes,
size 10 × 10, Holds 100 x cryotubes
Expand
View Pricing
EP0030140516
Storage Box for Eppendorf
®
Tubes,
size 9 × 9, 81 screw cap (cryo.) tubes 1-2 mL
Expand
View Pricing
EP0030140540
Storage Box for Eppendorf
®
Tubes,
size 9 × 9, Holds 81 x 3 mL tubes
Expand
View Pricing