Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Storage bottles without joints
Products
Product Category
bottles (4)
Brand
Sigma-Aldrich (4)
Manufacturer
Capacity (mL)
Cap
Bottle material
Color
Vial Bottle Material
Vial Bottle Color
Package Size
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
Z103284
Storage bottles without joints,
capacity 125 mL, PTFE stopcock
Expand
View Pricing
Z103292
Storage bottles without joints,
capacity 250 mL, PTFE stopcock
Expand
View Pricing
Z102482
Storage bottles without joints,
capacity 1,000 mL, PTFE stopcock
Expand
View Pricing
Z101990
Storage bottles without joints,
capacity 500 mL, PTFE stopcock
Expand
View Pricing