Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Storage and Distribution System
Products
Product Category
water storage and distribution systems (11)
Brand
Milli-Q (11)
Capacity (L)
Volume (ml)
Compatibility
Filter Products
Showing 1-11 of 11
Compare
Product Number
Description
Pricing
TANK6DP500
Storage and Distribution System,
duplex pump (100 V/120 V), Storage tank for pure (Type 2 and Type 3) water
Expand
View Pricing
TANK5DP500
Storage and Distribution System,
duplex pump (220 V/240 V), Storage tank for pure (Type 2 and Type 3) water
Expand
View Pricing
TANK5SP500
Storage and Distribution System,
single pump (220 V/240 V), Storage tank for pure (Type 2 and Type 3) water
Expand
View Pricing
TANK6DP500C
Storage and Distribution System,
duplex pump (120 V), sold only in US & Canada, Storage tank for pure (Type 2 and Type 3) water
Expand
View Pricing
TANK00200
Storage and Distribution System,
200 L storage tank (no relay) for AFS
®
systems, For optimal storage of Type 2 or Type 3 pure water
Expand
View Pricing
TANK60200
Storage and Distribution System,
200 L storage tank (120 V) for Elix
®
and RiOs
™
systems, For optimal storage of Type 2 or Type 3 pure water
Expand
View Pricing
TANK6SP500C
Storage and Distribution System,
single pump (120 V), sold only in US & Canada, Storage tank for pure (Type 2 and Type 3) water
Expand
View Pricing
TANK6SP500
Storage and Distribution System,
single pump (100 V/127 V), Storage tank for pure (Type 2 and Type 3) water
Expand
View Pricing
TANK5P200
Storage and Distribution System,
200 L storage tank (230 V) for Elix
®
and RiOs
™
systems, For optimal storage of Type 2 or Type 3 pure water
Expand
View Pricing
TANK6P200
Storage and Distribution System,
200 L storage tank (120 V) for Elix
®
and RiOs
™
systems, For optimal storage of Type 2 or Type 3 pure water
Expand
View Pricing
TANK50200
Storage and Distribution System,
200 L storage tank (230 V) for Elix
®
and RiOs
™
systems, For optimal storage of Type 2 or Type 3 pure water
Expand
View Pricing