Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Stainless steel - AISI 420
Products
Manufacturer
Physical Form
Filter Products
Showing 1-8 of 8
Compare
Product Number
Description
Pricing
GF93838720
Stainless steel - AISI 420,
sheet, Fe86.7%/Cr13.0%/C 0.3%, thickness 1.5 mm, size 200 × 200 mm
Expand
View Pricing
GF89694781
Stainless steel - AISI 420,
sheet, Fe86.7%/Cr13.0%/C 0.3%, thickness 2.0 mm, size 90 × 90 mm
Expand
View Pricing
GF41044845
Stainless steel - AISI 420,
sheet, Fe 86.7%/Cr13.0%/C 0.3%, thickness 0.5 mm, size 50 × 50 mm
Expand
View Pricing
GF79345389
Stainless steel - AISI 420,
sheet, thickness 0.5 mm, Fe86.7%/Cr13.0%/C 0.3%, size 200 × 200 mm
Expand
View Pricing
GF97377126
Stainless steel - AISI 420,
sheet, thickness 0.5 mm, Fe86.7%/Cr13.0%/C 0.3%, size 100 × 100 mm
Expand
View Pricing
GF32240135
Stainless steel - AISI 420,
sheet, Fe 86.7%/Cr13.0%/C 0.3%, thickness 1.5 mm, size 50 × 50 mm
Expand
View Pricing
GF33357326
Stainless steel - AISI 420,
sheet, Fe 86.7%/Cr13.0%/C 0.3%, thickness 1.5 mm, size 100 × 100 mm
Expand
View Pricing
GF49301210
Stainless steel - AISI 420,
sheet, Fe 86.7%/Cr13.0%/C 0.3%, thickness 2.0 mm, size 180 × 180 mm
Expand
View Pricing