Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Stainless Steel - AISI 321 alloy, FeCr18Ni9Ti
Products
Manufacturer
Physical Form
Filter Products
Showing 1-20 of 201
Page 1 of 11
Page 1 of 11
Compare
Product Number
Description
Pricing
GF74387694
Stainless Steel - AISI 321 alloy, FeCr18Ni9Ti,
1000mm tube, 1.60mm outside diameter, 0.55mm wall thickness, 0.5mm inside diameter, hard
Expand
View Pricing
GF32838677
Stainless Steel - AISI 321 alloy, FeCr18Ni9Ti,
200mm rod, 12.7mm diameter
Expand
View Pricing
GF74130993
Stainless Steel - AISI 321 alloy, FeCr18Ni9Ti,
foil, 50 x 50mm, 0.2mm thickness, annealed
Expand
View Pricing
GF35636362
Stainless Steel - AISI 321 alloy, FeCr18Ni9Ti,
foil, 25 x 25mm, 0.05mm thickness, annealed, light tight (lt)
Expand
View Pricing
GF08023520
Stainless Steel - AISI 321 alloy, FeCr18Ni9Ti,
500mm rod, 25.4mm diameter
Expand
View Pricing
GF33493439
Stainless Steel - AISI 321 alloy, FeCr18Ni9Ti,
500mm rod, 6.35mm diameter
Expand
View Pricing
GF09795527
Stainless Steel - AISI 321 alloy, FeCr18Ni9Ti,
1000mm tube, 10.0mm outside diameter, 1.5mm wall thickness, 7mm inside diameter, annealed
Expand
View Pricing
GF49341247
Stainless Steel - AISI 321 alloy, FeCr18Ni9Ti,
500mm tube, 1.60mm outside diameter, 0.55mm wall thickness, 0.5mm inside diameter, hard
Expand
View Pricing
GF57316518
Stainless Steel - AISI 321 alloy, FeCr18Ni9Ti,
foil, 50 x 50mm, 0.914mm thickness, annealed
Expand
View Pricing
GF83591141
Stainless Steel - AISI 321 alloy, FeCr18Ni9Ti,
foil, 25 x 25mm, 0.25mm thickness, annealed
Expand
View Pricing
GF33381198
Stainless Steel - AISI 321 alloy, FeCr18Ni9Ti,
foil, 8mm disks, 0.2mm thickness, annealed
Expand
View Pricing
GF06754620
Stainless Steel - AISI 321 alloy, FeCr18Ni9Ti,
foil, 100 x 100mm, 0.025mm thickness, annealed, light tight (lt)
Expand
View Pricing
GF25689752
Stainless Steel - AISI 321 alloy, FeCr18Ni9Ti,
foil, 50mm disks, 0.0125mm thickness, annealed
Expand
View Pricing
GF57440246
Stainless Steel - AISI 321 alloy, FeCr18Ni9Ti,
500mm rod, 12.7mm diameter
Expand
View Pricing
GF87484292
Stainless Steel - AISI 321 alloy, FeCr18Ni9Ti,
100mm rod, 12.7mm diameter
Expand
View Pricing
GF01904172
Stainless Steel - AISI 321 alloy, FeCr18Ni9Ti,
500mm tube, 10.0mm outside diameter, 1.5mm wall thickness, 7mm inside diameter, annealed
Expand
View Pricing
GF14177291
Stainless Steel - AISI 321 alloy, FeCr18Ni9Ti,
foil, 100 x 100mm, 0.5mm thickness, annealed
Expand
View Pricing
GF11582388
Stainless Steel - AISI 321 alloy, FeCr18Ni9Ti,
foil, 100 x 100mm, 0.25mm thickness, annealed
Expand
View Pricing
GF56035313
Stainless Steel - AISI 321 alloy, FeCr18Ni9Ti,
foil, 50 x 50mm, 0.0125mm thickness, annealed, light tight (lt)
Expand
View Pricing
GF18853331
Stainless Steel - AISI 321 alloy, FeCr18Ni9Ti,
foil, 50 x 50mm, 0.0125mm thickness, hard, light tight (lt)
Expand
View Pricing
Page 1 of 11
Page 1 of 11