Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Stainless Steel - 17-4PH alloy, FeCr17Ni4
Products
Manufacturer
Physical Form
Diameter (mm)
Length (m)
Length (mm)
Filter Products
Showing 1-8 of 8
Compare
Product Number
Description
Pricing
GF06845902
Stainless Steel - 17-4PH alloy, FeCr17Ni4,
100mm rod, 12.7mm diameter
Expand
View Pricing
GF58450877
Stainless Steel - 17-4PH alloy, FeCr17Ni4,
1000mm rod, 16mm diameter
Expand
View Pricing
GF40035002
Stainless Steel - 17-4PH alloy, FeCr17Ni4,
1000mm rod, 12.7mm diameter
Expand
View Pricing
GF56380451
Stainless Steel - 17-4PH alloy, FeCr17Ni4,
500mm rod, 12.7mm diameter
Expand
View Pricing
GF94370508
Stainless Steel - 17-4PH alloy, FeCr17Ni4,
500mm rod, 16mm diameter
Expand
View Pricing
GF37927176
Stainless Steel - 17-4PH alloy, FeCr17Ni4,
200mm rod, 12.7mm diameter
Expand
View Pricing
GF87852673
Stainless Steel - 17-4PH alloy, FeCr17Ni4,
100mm rod, 16mm diameter
Expand
View Pricing
GF05439113
Stainless Steel - 17-4PH alloy, FeCr17Ni4,
200mm rod, 16mm diameter
Expand
View Pricing