Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Stainless Steel 1/16 in. Capillary Tubing
Products
Product Category
tubing (24)
Brand
Supelco (10)
Color
Length (cm)
Length (m)
Color
Filter Products
Showing 1-20 of 24
Page 1 of 2
Page 1 of 2
Compare
Product Number
Description
Pricing
56718-U
Stainless Steel 1/16 in. Capillary Tubing,
L × O.D. × I.D. 1.0 m × 1/16 in. × 0.007 in.
Expand
View Pricing
56708
Stainless Steel 1/16 in. Capillary Tubing,
L × O.D. × I.D. 10 cm × 1/16 in. × 0.005 in.
Expand
View Pricing
56720-U
Stainless Steel 1/16 in. Capillary Tubing,
L × O.D. × I.D. 10 cm × 1/16 in. × 0.010 in.
Expand
View Pricing
56734-U
Stainless Steel 1/16 in. Capillary Tubing,
L × O.D. × I.D. 30 cm × 1/16 in. × 0.030 in.
Expand
View Pricing
56710-U
Stainless Steel 1/16 in. Capillary Tubing,
L × O.D. × I.D. 30 cm × 1/16 in. × 0.005 in.
Expand
View Pricing
56711
Stainless Steel 1/16 in. Capillary Tubing,
L × O.D. × I.D. 0.5 m × 1/16 in. × 0.005 in.
Expand
View Pricing
56731
Stainless Steel 1/16 in. Capillary Tubing,
L × O.D. × I.D. 5 cm × 1/16 in. × 0.030 in.
Expand
View Pricing
56714
Stainless Steel 1/16 in. Capillary Tubing,
L × O.D. × I.D. 10 cm × 1/16 in. × 0.007 in.
Expand
View Pricing
56712-U
Stainless Steel 1/16 in. Capillary Tubing,
L × O.D. × I.D. 1.0 m × 1/16 in. × 0.005 in.
Expand
View Pricing
56728
Stainless Steel 1/16 in. Capillary Tubing,
L × O.D. × I.D. 30 cm × 1/16 in. × 0.020 in.
Expand
View Pricing
56730-U
Stainless Steel 1/16 in. Capillary Tubing,
L × O.D. × I.D. 1.0 m × 1/16 in. × 0.020 in.
Expand
View Pricing
56736
Stainless Steel 1/16 in. Capillary Tubing,
L × O.D. × I.D. 1.0 m × 1/16 in. × 0.030 in.
Expand
View Pricing
56724
Stainless Steel 1/16 in. Capillary Tubing,
L × O.D. × I.D. 1.0 m × 1/16 in. × 0.010 in.
Expand
View Pricing
56719
Stainless Steel 1/16 in. Capillary Tubing,
L × O.D. × I.D. 5 cm × 1/16 in. × 0.010 in.
Expand
View Pricing
56732
Stainless Steel 1/16 in. Capillary Tubing,
L × O.D. × I.D. 10 cm × 1/16 in. × 0.030 in.
Expand
View Pricing
56713
Stainless Steel 1/16 in. Capillary Tubing,
L × O.D. × I.D. 5 cm × 1/16 in. × 0.007 in.
Expand
View Pricing
56707
Stainless Steel 1/16 in. Capillary Tubing,
L × O.D. × I.D. 5 cm × 1/16 in. × 0.005 in.
Expand
View Pricing
56738
Stainless Steel 1/16 in. Capillary Tubing,
L × O.D. × I.D. 10 cm × 1/16 in. × 0.040 in.
Expand
View Pricing
56716
Stainless Steel 1/16 in. Capillary Tubing,
L × O.D. × I.D. 30 cm × 1/16 in. × 0.007 in.
Expand
View Pricing
56737
Stainless Steel 1/16 in. Capillary Tubing,
L × O.D. × I.D. 5 cm × 1/16 in. × 0.040 in.
Expand
View Pricing
Page 1 of 2
Page 1 of 2