Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Spinbar
®
magnetic stirring fleas
Products
Product Category
stir bars (17)
Brand
Sigma-Aldrich (1)
Color
Manufacturer
Diameter (mm)
Length (mm)
Package Size
Filter Products
Showing 1-17 of 17
Compare
Product Number
Description
Pricing
Z283819
Spinbar
®
magnetic stirring fleas,
white, L 5 mm, diam. 2 mm
Expand
View Pricing
Z283800
Spinbar
®
magnetic stirring fleas,
white, diam. 2 mm, L 2 mm
Expand
View Pricing
Z283827
Spinbar
®
magnetic stirring fleas,
white, L × diam. 7 mm × 2 mm
Expand
View Pricing
Z283851
Spinbar
®
magnetic stirring fleas,
white, diam. 3 mm, L 8 mm
Expand
View Pricing
Z283886
Spinbar
®
magnetic stirring fleas,
white, diam. 3 mm, L 12.7 mm
Expand
View Pricing
Z118850
Spinbar
®
magnetic stirring fleas,
red, L 10 mm, diam. 3 mm
Expand
View Pricing
Z283878
Spinbar
®
magnetic stirring fleas,
white, diam. 3 mm, L 10 mm
Expand
View Pricing
Z283789
Spinbar
®
magnetic stirring fleas,
white, diam. 1.5 mm, L 8 mm
Expand
View Pricing
Z283843
Spinbar
®
magnetic stirring fleas,
white, diam. 3 mm, L 6.35 mm
Expand
View Pricing
Z283835
Spinbar
®
magnetic stirring fleas,
white, diam. 3 mm, L 3 mm
Expand
View Pricing
Z118834
Spinbar
®
magnetic stirring fleas,
yellow, diam. 2 mm, L 7 mm
Expand
View Pricing
Z118826
Spinbar
®
magnetic stirring fleas,
red, diam. 2 mm, L 7 mm
Expand
View Pricing
Z283797
Spinbar
®
magnetic stirring fleas,
white, diam. 1.5 mm, L 15 mm
Expand
View Pricing
Z118796
Spinbar
®
magnetic stirring fleas,
yellow, diam. 2 mm, L 5 mm
Expand
View Pricing
Z118818
Spinbar
®
magnetic stirring fleas,
blue, diam. 2 mm, L 5 mm
Expand
View Pricing
Z118842
Spinbar
®
magnetic stirring fleas,
blue, diam. 2 mm, L 7 mm
Expand
View Pricing
Z118761
Spinbar
®
magnetic stirring fleas,
blue, diam. 1.5 mm, L 8 mm
Expand
View Pricing