Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Specialty Glass Column
Products
Product Category
columns (4)
Brand
Supelco (4)
ID (mm)
Length (mm)
OD (mm)
Quality Segment
Technique
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
64752
Specialty Glass Column,
glass coarse frit, PTFE stopcock, column L × O.D. × I.D. 300 mm × 13 mm × 10.5 mm, 415mm overall
Expand
View Pricing
64755
Specialty Glass Column,
glass coarse frit, PTFE stopcock, column L × O.D. × I.D. 400 mm × 25 mm × 22 mm, 515mm overall
Expand
View Pricing
64748
Specialty Glass Column,
reservoir design
, column L × O.D. × I.D. 200 mm × 9 mm × 7 mm, 280mm overall
Expand
View Pricing
64756
Specialty Glass Column,
glass coarse frit, PTFE stopcock, inlet joint: SJ, column L × O.D. × I.D. 300 mm × 13 mm × 10.5 mm, 415mm overall
Expand
View Pricing