Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
SP Bel-Art
®
Spinbar
®
magnetic stir bar
Synonym(s):
Spinbar
®
magnetic stir bar
Products
Product Category
stir bars (32)
Manufacturer
Diameter (mm)
Length (in)
Color
Package Size
Filter Products
Showing 1-20 of 32
Page 1 of 2
Page 1 of 2
Compare
Product Number
Description
Pricing
BAF371201060
SP Bel-Art
®
Spinbar
®
magnetic stir bar,
polygon white PTFE-coated (no pivot ring), L × diam. 2.375 in. (60 mm) × 0.375 in. (10 mm)
Expand
View Pricing
BAF371220050
SP Bel-Art
®
Spinbar
®
magnetic stir bar,
polygon white PTFE-coated, L × diam. 2 in. (50 mm) × 0.3125 in. (8 mm)
Expand
View Pricing
BAF371201276
SP Bel-Art
®
Spinbar
®
magnetic stir bar,
round white PTFE-coated (no pivot ring), L × diam. 3 in. (76 mm) × 0.5 in. (12 mm)
Expand
View Pricing
BAF371220060
SP Bel-Art
®
Spinbar
®
magnetic stir bar,
polygon white PTFE-coated, L × diam. 2.375 in. (60 mm) × 0.375 in. (10 mm)
Expand
View Pricing
BAF371111128
SP Bel-Art
®
Spinbar
®
magnetic stir bar,
cylindrical white PTFE-coated, L × diam. 1.5 in. (38.1 mm) × 0.375 in. (9.5 mm)
Expand
View Pricing
BAF371090020
SP Bel-Art
®
Spinbar
®
magnetic stir bar,
octagonal yellow PTFE-coated, L × diam. 1.5 in. (38.1 mm) × 0.3125 in. (8 mm)
Expand
View Pricing
BAF371220070
SP Bel-Art
®
Spinbar
®
magnetic stir bar,
polygon white PTFE-coated, L × diam. 2.75 in. (70 mm) × 0.375 in. (10 mm)
Expand
View Pricing
BAF371220025
SP Bel-Art
®
Spinbar
®
magnetic stir bar,
polygon white PTFE-coated, L × diam. 1 in. (25 mm) × 0.25 in. (6 mm)
Expand
View Pricing
BAF371200080
SP Bel-Art
®
Spinbar
®
magnetic stir bar,
polygon white PTFE-coated (no pivot ring), L × diam. 3.125 in. (80 mm) × 0.375 in. (10 mm)
Expand
View Pricing
BAF371220020
SP Bel-Art
®
Spinbar
®
magnetic stir bar,
polygon white PTFE-coated, L × diam. 0.75 in. (20 mm) × 0.25 in. (6 mm)
Expand
View Pricing
BAF371110138
SP Bel-Art
®
Spinbar
®
magnetic stir bar,
cylindrical white PTFE-coated, L × diam. 1 in. (25.4 mm) × 0.375 in. (9.5 mm)
Expand
View Pricing
BAF371090034
SP Bel-Art
®
Spinbar
®
magnetic stir bar,
octagonal red PTFE-coated, L × diam. 12.7 in. × 3.2 in.
Expand
View Pricing
BAF371090033
SP Bel-Art
®
Spinbar
®
magnetic stir bar,
octagonal blue PTFE-coated, L × diam. 3 in. (76.2 mm) × 0.5 in. (12.7 mm)
Expand
View Pricing
BAF371300158
SP Bel-Art
®
Spinbar
®
magnetic stir bar,
egg-shaped white PTFE-coated, L × diam. 1.625 in. (41.3 mm) × 0.75 (19)
Expand
View Pricing
BAF371220035
SP Bel-Art
®
Spinbar
®
magnetic stir bar,
polygon white PTFE-coated, L × diam. 1.375 in. (35 mm) × 0.25 in. (6 mm)
Expand
View Pricing
BAF371090036
SP Bel-Art
®
Spinbar
®
magnetic stir bar,
octagonal blue PTFE-coated, L × diam. 12.7 in. × 3.2 in.
Expand
View Pricing
BAF371090032
SP Bel-Art
®
Spinbar
®
magnetic stir bar,
octagonal yellow PTFE-coated, L × diam. 3 in. (76.2 mm) × 0.5 in. (12.7 mm)
Expand
View Pricing
BAF371110114
SP Bel-Art
®
Spinbar
®
magnetic stir bar,
cylindrical white PTFE-coated, L × diam. 1.25 in. (31.8 mm) × 0.3125 in. (8 mm)
Expand
View Pricing
BAF371200010
SP Bel-Art
®
Spinbar
®
magnetic stir bar,
polygon white PTFE-coated (no pivot ring), L × diam. 0.375 in. (10 mm) × 0.25 in. (6 mm)
Expand
View Pricing
BAF371090030
SP Bel-Art
®
Spinbar
®
magnetic stir bar,
octagonal blue PTFE-coated, L × diam. 2 in. (50.8 mm) × 0.3125 in. (8 mm)
Expand
View Pricing
Page 1 of 2
Page 1 of 2