Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
SP Bel-Art
®
Sampling Scoop
Products
Product Category
spatulas/scoops (4)
Manufacturer
Capacity (mL)
Height (mm)
Length (in)
Length (mm)
Color
Package Size
Sterility
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
BAF369330021
SP Bel-Art
®
Sampling Scoop,
white polystyrene, bowl capacity 63 mL (2 oz), pack of 12 ea
Expand
View Pricing
BAF369330022
SP Bel-Art
®
Sampling Scoop,
white polystyrene, bowl capacity 83 mL (2.7 oz), pack of 12 ea
Expand
View Pricing
BAF369330024
SP Bel-Art
®
Sampling Scoop,
white polystyrene, bowl capacity 250 mL (8 oz) , 8 oz, pack of 12 ea
Expand
View Pricing
BAF369330023
SP Bel-Art
®
Sampling Scoop,
white polystyrene, bowl capacity 125 mL (4 oz), pack of 12 ea
Expand
View Pricing