Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
SP Bel-Art
®
H-B FRIO-Temp
®
Freezer Verification Thermometer
Products
Manufacturer
Length (in)
Volume (ml)
Package Size
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
BAB610040300
SP Bel-Art
®
H-B FRIO-Temp
®
Freezer Verification Thermometer,
temp -30-1 °C, Thermometer fill, Enviro-Safe
®
Liquid, bottle volume 30 mL, Bottle fill, Enviro-Safe
®
Liquid
Expand
View Pricing
BAB610010100
SP Bel-Art
®
H-B FRIO-Temp
®
Freezer Verification Thermometer,
temp -25--5 °C, bottle volume 60 mL, Bottle fill, Enviro-Safe
®
Liquid
Expand
View Pricing
BAB610040900
SP Bel-Art
®
H-B FRIO-Temp
®
Freezer Verification Thermometer,
temp -20-35 °C, bottle volume 30 mL, Thermometer fill, Enviro-Safe
®
Liquid, Bottle fill, Enviro-Safe
®
Liquid
Expand
View Pricing
BAB602000700
SP Bel-Art
®
H-B FRIO-Temp
®
Freezer Verification Thermometer,
temp -30-1 °C
Expand
View Pricing