Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
SP Bel-Art
®
H-B Enviro-Safe
®
Enviro-Safe Liquid-In-Glass Pocket Thermometer
Synonym(s):
Enviro-Safe
®
pocket thermometer
Products
Manufacturer
Length (in)
Length (mm)
Package Size
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
BAB605700300
SP Bel-Art
®
H-B Enviro-Safe
®
Enviro-Safe Liquid-In-Glass Pocket Thermometer,
temp 0-220 °F, plastic case (with Window )
Expand
View Pricing
BAB605700100
SP Bel-Art
®
H-B Enviro-Safe
®
Enviro-Safe Liquid-In-Glass Pocket Thermometer,
temp -10-110 °C, plastic case (with Window )
Expand
View Pricing
BAB605701000
SP Bel-Art
®
H-B Enviro-Safe
®
Enviro-Safe Liquid-In-Glass Pocket Thermometer,
temp -5-50 °C, metal case (with Window )
Expand
View Pricing
BAB605700200
SP Bel-Art
®
H-B Enviro-Safe
®
Enviro-Safe Liquid-In-Glass Pocket Thermometer,
temp -5-50 °C, plastic case (with Window )
Expand
View Pricing
BAB605700400
SP Bel-Art
®
H-B Enviro-Safe
®
Enviro-Safe Liquid-In-Glass Pocket Thermometer,
temp 20-120 °C, plastic case (with Window )
Expand
View Pricing