Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
S-Pak
®
Membrane Filter
Products
Product Category
filters (10)
Brand
Millipore (10)
Agency
Diameter (mm)
Compatibility
Hydrophobicity
Application
Color
Pore size (µm)
Sterility
Sterilization Method
Filter Products
Showing 1-10 of 10
Compare
Product Number
Description
Pricing
HAWG047S6
S-Pak
®
Membrane Filter,
mixed cellulose esters (MCE), pore size 0.45 μm, filter diam. 47 mm, white gridded membranes for microbiological analysis
Expand
View Pricing
HABG047S6
S-Pak
®
Membrane Filter,
mixed cellulose esters (MCE), pore size 0.45 μm, filter diam. 47 mm, black gridded membranes for microbiological analysis
Expand
View Pricing
GSWG047S6
S-Pak
®
Membrane Filter,
mixed cellulose esters (MCE), pore size 0.22 μm, filter diam. 47 mm, white gridded membranes for microbiological analysis
Expand
View Pricing
HVWP047S6
S-Pak
®
Membrane Filter,
PVDF, pore size 0.45 μm, filter diam. 47 mm, white plain sterile membranes for microbiological analysis
Expand
View Pricing
HCWG047S6
S-Pak
®
Membrane Filter,
mixed cellulose esters (MCE), pore size 0.7 μm, filter diam. 47 mm, white gridded membranes for microbiological analysis of wastewater
Expand
View Pricing
RAWG047S6
S-Pak
®
Membrane Filter,
mixed cellulose esters (MCE), pore size 1.2 μm, filter diam. 47 mm, white gridded sterile membranes for microbiological analysis
Expand
View Pricing
AAWG047S6
S-Pak
®
Membrane Filter,
mixed cellulose esters (MCE), pore size 0.8 μm, filter diam. 47 mm, white gridded membranes for microbiological analysis of beverages
Expand
View Pricing
HAWG647SP
S-Pak
®
Membrane Filter,
mixed cellulose esters (MCE), pore size 0.45 μm, filter diam. 47 mm, white gridded + 3 dispensers & 600pads for microbiological analysis
Expand
View Pricing
HAGG050S6
S-Pak
®
Membrane Filter,
mixed cellulose esters (MCE), pore size 0.45 μm, filter diam. 50 mm, green gridded for general microbiological analysis
Expand
View Pricing
HAWG050S6
S-Pak
®
Membrane Filter,
mixed cellulose esters (MCE), pore size 0.45 μm, filter diam. 50 mm, white gridded for general microbiological analysis
Expand
View Pricing