Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Space Saver storage racks
Products
Product Category
racks (5)
Brand
Supelco (5)
Height (mm)
Length (mm)
Color
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
23207
Space Saver storage racks,
for use with 11-12 mm diameter vial (50 vials per rack), well opening 12.5 mm, pkg of 5 ea
Expand
View Pricing
23202
Space Saver storage racks,
for use with 17 mm diameter vial (90 vials per rack), well opening 17.1 mm, pkg of 5 ea
Expand
View Pricing
23201
Space Saver storage racks,
for use with 21-23 mm diameter vial (36 vials per rack), well opening 23.1 mm, pkg of 5 ea
Expand
View Pricing
23206
Space Saver storage racks,
for use with 29 mm diameter vial (50 vials per rack), well opening 30.0 mm, pkg of 5 ea
Expand
View Pricing
23205-U
Space Saver storage racks,
for use with 15-16 mm diameter vial (48 vials per rack), well opening 15.5 mm, pkg of 5 ea
Expand
View Pricing