Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Sodium succinate dibasic hexahydrate
Synonym(s):
Butanedioic acid disodium salt, Succinic acid disodium salt
Linear Formula:
NaOOCCH
2
CH
2
COONa · 6H
2
O
CAS Number:
6106-21-4
Molecular Weight:
270.14
EC Number:
205-778-7
Products
Brand
Sigma-Aldrich (3)
Formula Weight
Physical Form
Product Line
Purity
Research Area
Quality Segment
Markush Class
Markush Group
Special Grade
Puriss (1)
Puriss P.A. (1)
Technique
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
S2378
Sodium succinate dibasic hexahydrate,
ReagentPlus
®
, ≥99%
Expand
View Pricing
S9637
Sodium succinate dibasic hexahydrate,
BioReagent, suitable for cell culture, suitable for insect cell culture
Expand
View Pricing
14170
Sodium succinate dibasic hexahydrate,
puriss. p.a., ≥98.0% (NT)
Expand
View Pricing