Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Sodium pyrophosphate dibasic
Synonym(s):
Disodium dihydrogen pyrophosphate, Sodium diphosphate dibasic
Linear Formula:
Na
2
H
2
P
2
O
7
CAS Number:
7758-16-9
Molecular Weight:
221.94
EC Number:
231-835-0
Products
Product Category
buffers (1)
salts (1)
Formula Weight
pH Value
Physical Form
Product Line
Purity
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
71501
Sodium pyrophosphate dibasic,
BioUltra, ≥99.0% (T)
Expand
View Pricing
P8135
Sodium pyrophosphate dibasic,
practical grade
Expand
View Pricing