Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Sodium nitrate-
15
N
Linear Formula:
Na
15
NO
3
CAS Number:
31432-45-8
Molecular Weight:
85.99
Products
Product Category
salts (5)
stable isotopes (5)
Brand
Sigma-Aldrich (5)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Isotope
Application
Markush Class
Markush Group
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
364606
Sodium nitrate-
15
N,
≥98 atom %
15
N, ≥99% (CP)
Expand
View Pricing
490806
Sodium nitrate-
15
N,
60 atom %
15
N
Expand
View Pricing
490792
Sodium nitrate-
15
N,
10 atom %
15
N
Expand
View Pricing
490784
Sodium nitrate-
15
N,
5 atom %
15
N
Expand
View Pricing
576603
Sodium nitrate-
15
N,
25-30 atom %
15
N
Expand
View Pricing