Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Sodium molybdate
Linear Formula:
Na
2
MoO
4
CAS Number:
7631-95-0
Molecular Weight:
205.92
EC Number:
231-551-7
Products
Product Category
salts (1)
Brand
Sigma-Aldrich (2)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Application
Reagent Type
Core
Special Grade
Anhydrous (1)
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
243655
Sodium molybdate,
≥98%
Expand
View Pricing
737860
Sodium molybdate,
anhydrous, powder, −100 mesh particle size, 99.9% trace metals basis
Expand
View Pricing