Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Sodium hydrogen carbonate
Synonym(s):
Sodium bicarbonate
Linear Formula:
NaHCO
3
CAS Number:
144-55-8
Molecular Weight:
84.01
Products
Product Category
salts (4)
bases (1)
buffers (1)
Brand
SAFC (3)
Sigma-Aldrich (1)
Formula Weight
Physical Form
pK Value
Product Line
Purity
Agency
Quality Segment
Application
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
1.06329
Sodium hydrogen carbonate,
for analysis EMSURE
®
ACS,Reag.Ph Eur
Expand
View Pricing
1.37013
Sodium hydrogen carbonate,
EMPROVE
®
EXPERT, Ph. Eur., BP, ChP, JP, USP
Expand
View Pricing
1.06323
Sodium hydrogen carbonate,
EMPROVE
®
ESSENTIAL, Ph. Eur., BP, ChP, JP, USP, FCC, E 500
Expand
View Pricing
401676
Sodium hydrogen carbonate,
−40-+140 mesh, ≥95%
Expand
View Pricing
1.72577
Sodium hydrogen carbonate,
for table water, FCC, E 500
Expand
View Pricing