Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Sodium chloride solution
Linear Formula:
NaCl
CAS Number:
7647-14-5
Molecular Weight:
58.44
Products
Product Category
salts (6)
buffers (1)
Brand
SAFC (2)
Sigma-Aldrich (4)
Feature
Formula Weight
pH Value
Physical Form
Product Line
Research Area
Quality Segment
Application
Special Grade
GMO free (1)
GMP (1)
Halal (1)
Kosher (1)
Technical (1)
Sterility
Sterilization Method
Technique
Filter Products
Showing 1-8 of 8
Compare
Product Number
Description
Pricing
S5150
Sodium chloride solution,
5 M in H
2
O, BioReagent, for molecular biology, suitable for cell culture
Expand
View Pricing
S8776
Sodium chloride solution,
0.9% in water, BioXtra, suitable for cell culture
Expand
View Pricing
59222C
Sodium chloride solution,
5 M
Expand
View Pricing
71386
Sodium chloride solution,
BioUltra, for molecular biology, ~5 M in H
2
O
Expand
View Pricing
71392
Sodium chloride solution,
technical, ~26% (saturated in water at 20°C, AT)
Expand
View Pricing
S6546
Sodium chloride solution,
5 M
Expand
View Pricing
1.37076
Sodium chloride solution,
3 mol/l, EMPROVE
®
EXPERT
Expand
View Pricing
S0817
Sodium chloride solution,
0.85%
Expand
View Pricing