Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Sodium cacodylate trihydrate
Synonym(s):
Cacodylic acid sodium salt trihydrate, Dimethylarsinic acid sodium salt, Dimethylarsonic acid sodium salt
Linear Formula:
(CH
3
)
2
AsO
2
Na · 3H
2
O
CAS Number:
6131-99-3
Molecular Weight:
214.03
EC Number:
204-708-2
Products
Product Category
buffers (2)
Brand
Sigma-Aldrich (3)
Feature
Formula Weight
Physical Form
Product Line
Purity
Quality Segment
Application
Special Grade
Purum (1)
Purum P.A. (1)
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
C0250
Sodium cacodylate trihydrate,
≥98%
Expand
View Pricing
C4945
Sodium cacodylate trihydrate,
BioXtra, ≥98%
Expand
View Pricing
20840
Sodium cacodylate trihydrate,
purum p.a., ≥98.0% (T)
Expand
View Pricing