Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Slide box
Products
Product Category
boxes (6)
Color
Manufacturer
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
HS15991A
Slide box,
100 place, blue
Expand
View Pricing
HS15991C
Slide box,
100 place, white
Expand
View Pricing
HS15991B
Slide box,
100 place, red
Expand
View Pricing
HS15990A
Slide box,
25 place, blue
Expand
View Pricing
HS15990C
Slide box,
25 place, white
Expand
View Pricing
HS15990B
Slide box,
25 place, red
Expand
View Pricing