Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Silver-copper alloy, Ag72Cu28
Products
Manufacturer
Physical Form
Diameter (mm)
Length (m)
Length (mm)
Filter Products
Showing 1-20 of 34
Page 1 of 2
Page 1 of 2
Compare
Product Number
Description
Pricing
GF89551389
Silver-copper alloy, Ag72Cu28,
foil, 50 x 50mm, 0.25mm thickness
Expand
View Pricing
GF92851162
Silver-copper alloy, Ag72Cu28,
powder, 45micron max. particle size, 20g
Expand
View Pricing
GF83824083
Silver-copper alloy, Ag72Cu28,
foil, 100 x 100mm, 0.25mm thickness
Expand
View Pricing
GF07146525
Silver-copper alloy, Ag72Cu28,
100m wire, 0.25mm diameter, as drawn
Expand
View Pricing
GF21808661
Silver-copper alloy, Ag72Cu28,
20m wire, 0.8mm diameter, as drawn
Expand
View Pricing
GF99418898
Silver-copper alloy, Ag72Cu28,
powder, 45micron max. particle size, 100g
Expand
View Pricing
GF06395351
Silver-copper alloy, Ag72Cu28,
powder, 45micron max. particle size, 50g
Expand
View Pricing
GF19011844
Silver-copper alloy, Ag72Cu28,
foil, 50 x 50mm, 2.0mm thickness
Expand
View Pricing
GF62662675
Silver-copper alloy, Ag72Cu28,
200mm rod, 2.0mm diameter, as drawn
Expand
View Pricing
GF47279065
Silver-copper alloy, Ag72Cu28,
foil, 50 x 50mm, 0.05mm thickness, light tight (lt)
Expand
View Pricing
GF40346669
Silver-copper alloy, Ag72Cu28,
500mm rod, 2.0mm diameter, as drawn
Expand
View Pricing
GF54445215
Silver-copper alloy, Ag72Cu28,
100mm rod, 2.0mm diameter, as drawn
Expand
View Pricing
GF70377868
Silver-copper alloy, Ag72Cu28,
5m wire, 0.5mm diameter, as drawn
Expand
View Pricing
GF33493743
Silver-copper alloy, Ag72Cu28,
10m wire, 0.25mm diameter, as drawn
Expand
View Pricing
GF97061081
Silver-copper alloy, Ag72Cu28,
foil, 100 x 100mm, 0.05mm thickness, light tight (lt)
Expand
View Pricing
GF61993581
Silver-copper alloy, Ag72Cu28,
1000mm rod, 2.0mm diameter, as drawn
Expand
View Pricing
GF58590771
Silver-copper alloy, Ag72Cu28,
5m wire, 0.25mm diameter, as drawn
Expand
View Pricing
GF20633655
Silver-copper alloy, Ag72Cu28,
foil, 0.5m coil, 0.05mm thickness
Expand
View Pricing
GF45667592
Silver-copper alloy, Ag72Cu28,
100mm rod, 12.5mm diameter, as drawn
Expand
View Pricing
GF38958980
Silver-copper alloy, Ag72Cu28,
20m wire, 0.5mm diameter, as drawn
Expand
View Pricing
Page 1 of 2
Page 1 of 2